
Ethylene glycol monobutyl ether

Synonym(s):
Ethylene glycol butyl ether, 2-Butoxyethanol, Butyl glycol
Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1