Ethinyl estradiol Related Compound A
Synonym(s):
(17α)-3,17-Dihydroxy-19-norpregna-1,3,5(10)-trien-20-yn-6-one, 3,17β-Dihydroxy-17α-ethynylestra-1,3,5(10)-trien-6-one, 6-Keto-ethinyl estradiol, 6-Oxo-17-α-ethynylestradiol
Empirical Formula (Hill Notation):
C20H22O3
CAS Number:
Molecular Weight:
310.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing