Skip to Content
Merck
vibrant-m

Eszopiclone Related Compound A

vibrant-m
Synonym(s):
Zopiclone oxide, (5RS)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate 4-oxide
Empirical Formula (Hill Notation):
C17H17ClN6O4
CAS Number:
Molecular Weight:
404.81

Products

Brand
Formula Weight
Agency
Quality Segment
Application
API Family
Special Grade
Showing 1-1 of 1