![Epirubicin hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/123/880/ed0f3881-8460-43fb-b003-35f88731f853/640/ed0f3881-8460-43fb-b003-35f88731f853.png)
Epirubicin hydrochloride
![Epirubicin hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/123/880/ed0f3881-8460-43fb-b003-35f88731f853/640/ed0f3881-8460-43fb-b003-35f88731f853.png)
Synonym(s):
4′-Epidoxorubicin hydrochloride, Epidoxorubicin hydrochloride
Empirical Formula (Hill Notation):
C27H29NO11 · HCl
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing