Skip to Content
Merck
Ebastine European Pharmacopoeia (EP) Reference Standard

Ebastine

Ebastine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-piperidinyl]-1-butanone
Empirical Formula (Hill Notation):
C32H39NO2
CAS Number:
Molecular Weight:
469.66

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing