Skip to Content
Merck
Dysprosium rod, 50mm, diameter 2.0mm, cast, 99%

Dysprosium

Dysprosium rod, 50mm, diameter 2.0mm, cast, 99%
Synonym(s):
Dysprosium
Empirical Formula (Hill Notation):
Dy
CAS Number:
Molecular Weight:
162.50

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Showing 1-20 of 25
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2