![Dronedarone hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/417/878/2ddfdb99-231a-4cec-9ba5-c1a67600950d/640/2ddfdb99-231a-4cec-9ba5-c1a67600950d.png)
Dronedarone hydrochloride
![Dronedarone hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/417/878/2ddfdb99-231a-4cec-9ba5-c1a67600950d/640/2ddfdb99-231a-4cec-9ba5-c1a67600950d.png)
Synonym(s):
N-{2-Butyl-3-[4-(3-dibutylaminopropoxy)benzoyl]benzofuran-5-yl}methanesulfonamide hydrochloride, Multaq, SR-33589B
Empirical Formula (Hill Notation):
C31H44N2O5S · HCl
CAS Number:
Molecular Weight:
593.22
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3