
Doxepin Related Compound B

Synonym(s):
(11RS)-11-[3-(Dimethylamino)propyl]-6,11-dihydrodibenz[b,e]oxepin-11-ol
Empirical Formula (Hill Notation):
C19H23NO2
CAS Number:
Molecular Weight:
297.39
Products
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing