Skip to Content
Merck
Doxazosin Related Compound B certified reference material, pharmaceutical secondary standard

Doxazosin Related Compound B

Doxazosin Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
6,7-Dimethoxy-2,4-quinazolinedione
Empirical Formula (Hill Notation):
C10H10N2O4
CAS Number:
Molecular Weight:
222.20

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade