Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DOTMA
Empirical Formula (Hill Notation):
C
42
H
84
ClNO
2
CAS Number:
104872-42-6
Molecular Weight:
670.57
Products
Product Category
complex lipids (2)
lipids (2)
Formula Weight
Manufacturer
Physical Form
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
890898P
DOTMA,
Avanti Research
™
- A Croda Brand
Expand
View Pricing
890898C
DOTMA,
Avanti Research
™
- A Croda Brand
Expand
View Pricing