![DMT-dG(ib) Pharmadite®](/deepweb/assets/sigmaaldrich/product/structures/373/838/b5e54924-c87e-404e-b87e-3de0ccee0ede/640/b5e54924-c87e-404e-b87e-3de0ccee0ede.png)
DMT-dG(ib) Pharmadite®
![DMT-dG(ib) Pharmadite®](/deepweb/assets/sigmaaldrich/product/structures/373/838/b5e54924-c87e-404e-b87e-3de0ccee0ede/640/b5e54924-c87e-404e-b87e-3de0ccee0ede.png)
Synonym(s):
DMT-dG(ib) Phosphoramidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C44H54N7O8P
CAS Number:
Molecular Weight:
839.92
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing