Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-Mannoheptulose
Synonym(s):
D-Manno-2-heptulose
Empirical Formula (Hill Notation):
C
7
H
14
O
7
CAS Number:
3615-44-9
Molecular Weight:
210.18
EC Number:
222-795-5
Products
Product Category
carbohydrates (1)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
97318
D-Mannoheptulose,
≥99.0% (HPLC)
Expand
View Pricing