Skip to Content
Merck
(±)-Norverapamil hydrochloride ≥98% (HPLC)

(±)-Norverapamil hydrochloride

(±)-Norverapamil hydrochloride ≥98% (HPLC)
Synonym(s):
N-Nor-(±)-verapamil hydrochloride
Empirical Formula (Hill Notation):
C26H36N2O4 · HCl
CAS Number:
Molecular Weight:
477.04

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing