![(±)-Nicotine-(pyridine-d4) ≥98 atom % D, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/534/352/7045eead-c113-4e86-934a-0de331842577/640/7045eead-c113-4e86-934a-0de331842577.png)
(±)-Nicotine-(pyridine-d4)
![(±)-Nicotine-(pyridine-d4) ≥98 atom % D, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/534/352/7045eead-c113-4e86-934a-0de331842577/640/7045eead-c113-4e86-934a-0de331842577.png)
Empirical Formula (Hill Notation):
C10D4H10N2
CAS Number:
Molecular Weight:
166.26
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing