![D,L-Camphene phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/266/562/8778743a-4448-48e4-8ce0-c0532337ffbd/640/8778743a-4448-48e4-8ce0-c0532337ffbd.png)
D,L-Camphene
![D,L-Camphene phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/266/562/8778743a-4448-48e4-8ce0-c0532337ffbd/640/8778743a-4448-48e4-8ce0-c0532337ffbd.png)
Synonym(s):
Camphene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing