Skip to Content
Merck
DL-Alanine ≥99%, FCC, FG

DL-Alanine

DL-Alanine ≥99%, FCC, FG
Synonym(s):
(±)-2-Aminopropionic acid
Linear Formula:
CH3CH(NH2)COOH
CAS Number:
Molecular Weight:
89.09
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Amino Acid
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing