Skip to Content
Merck
di-Sodium hydrogen phosphate dihydrate suitable for HPLC, LiChropur™

di-Sodium hydrogen phosphate dihydrate

di-Sodium hydrogen phosphate dihydrate suitable for HPLC, LiChropur™
Synonym(s):
sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate, di-Sodium hydrogen phosphate dihydrate
Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique