Diphenyl phosphoryl azide
Synonym(s):
DPPA, Phosphoric acid diphenyl ester azide
Linear Formula:
(C6H5O)2P(O)N3
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1