Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dimethyldioctadecylammonium bromide
Synonym(s):
Distearyldimethylammonium bromide
Linear Formula:
[CH
3
(CH
2
)
17
]
2
N(Br)(CH
3
)
2
CAS Number:
3700-67-2
Molecular Weight:
630.95
EC Number:
223-037-6
Products
Product Category
detergents (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D2779
Dimethyldioctadecylammonium bromide,
≥98% (TLC)
Expand
View Pricing
40165
Dimethyldioctadecylammonium bromide,
≥98.0% (AT)
Expand
View Pricing