Skip to Content
Merck
Diclofenac sodium British Pharmacopoeia (BP) Reference Standard

Diclofenac sodium

Diclofenac sodium British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
EC Number:

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing