Skip to Content
Merck
Diatrizoic Acid Related Compound A certified reference material, pharmaceutical secondary standard

Diatrizoic Acid Related Compound A

Diatrizoic Acid Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
3-Acetamido-5-amino-2,4,6-triiodobenzoic acid, 3-(Acetylamino)-5-amino-2,4,6-triiodobenzoic acid
Empirical Formula (Hill Notation):
C9H7I3N2O3
CAS Number:
Molecular Weight:
571.88

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade