![D-Fructose 1,6-bisphosphate trisodium salt hydrate ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/338/716/8c953323-0b16-44a6-b2e7-4a61ca050ba1/640/8c953323-0b16-44a6-b2e7-4a61ca050ba1.png)
D-Fructose 1,6-bisphosphate trisodium salt hydrate
![D-Fructose 1,6-bisphosphate trisodium salt hydrate ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/338/716/8c953323-0b16-44a6-b2e7-4a61ca050ba1/640/8c953323-0b16-44a6-b2e7-4a61ca050ba1.png)
Synonym(s):
D(+)Fructofuranose 1,6-diphosphate trisodium salt hydrate, Harden-Young ester, Hexose diphosphate trisodium salt hydrate
Empirical Formula (Hill Notation):
C6H11Na3O12P2 · xH2O
CAS Number:
Molecular Weight:
406.06 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing