Dextroamphetamine Related Compound B
Synonym(s):
Phenylacetone, Benzyl methyl ketone, Phenyl-2-propanone
Linear Formula:
C6H5CH2COCH3
CAS Number:
Molecular Weight:
134.18
Products
Brand
Feature
Formula Weight
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing