![Dehydrocostus lactone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/814/514/faf1f144-2086-41e1-9ba9-2fe64ae28625/640/faf1f144-2086-41e1-9ba9-2fe64ae28625.png)
Dehydrocostus lactone
![Dehydrocostus lactone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/814/514/faf1f144-2086-41e1-9ba9-2fe64ae28625/640/faf1f144-2086-41e1-9ba9-2fe64ae28625.png)
Synonym(s):
(−)-Dehydrocostus lactone; Dehydrocostus lactone; Epiligulyl oxide, (3aS,6aR,9aR,9bS)-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one, Dehydro-costus lactone (6CI,7CI), Dehydrocostuslactone, Guaia-4(15),10(14),11(13)-trien-12-oic acid, 6alpha-hydroxy-, gamma-lactone, [3aS-(3aα,6aα,9aα,9bβ)]-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one
Empirical Formula (Hill Notation):
C15H18O2
CAS Number:
Molecular Weight:
230.30
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing