Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Decamethonium bromide
Synonym(s):
Decamethylene bis(trimethylammonium bromide), Decane-1,10-bis(trimethylammonium bromide)
Linear Formula:
(CH
3
)
3
N(Br)(CH
2
)
10
N(Br)(CH
3
)
3
CAS Number:
541-22-0
Molecular Weight:
418.29
EC Number:
208-772-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D1260
Decamethonium bromide,
crystalline
Expand
View Pricing
396966
Decamethonium bromide,
Ion-Pair reagent, ≥99%
Expand
View Pricing