Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(+)-α-Tocopherol
Synonym(s):
2,5,7,8-Tetramethyl-2-(4′,8′,12′-trimethyltridecyl)-6-chromanol, 5,7,8-Trimethyltocol, D-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C
29
H
50
O
2
CAS Number:
59-02-9
Molecular Weight:
430.71
EC Number:
200-412-2
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Color
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T3634
(+)-α-Tocopherol,
from vegetable oil, Type V, ~1000 IU/g
Expand
View Pricing
T1539
(+)-α-Tocopherol,
Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Expand
View Pricing