Skip to Content
Merck
DAPH ≥98% (HPLC), solid

DAPH

DAPH ≥98% (HPLC), solid
Synonym(s):
4,5-Dianilinophthalimide, 5,6-Bis(phenylamino)-1H-isoindole-1,3(2H)-dione, CGP 52411
Empirical Formula (Hill Notation):
C20H15N3O2
CAS Number:
Molecular Weight:
329.35

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing