![Cyanazine PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/107/296/cbc9af79-d04e-4edd-8e9c-1154a684ec45/640/cbc9af79-d04e-4edd-8e9c-1154a684ec45.png)
Cyanazine
![Cyanazine PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/107/296/cbc9af79-d04e-4edd-8e9c-1154a684ec45/640/cbc9af79-d04e-4edd-8e9c-1154a684ec45.png)
Empirical Formula (Hill Notation):
C9H13ClN6
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing