Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cortisone 21-acetate
Synonym(s):
17α,21-Dihydroxy-4-pregnene-3,11,20-trione 21-acetate, 21-Acetoxy-4-pregnen-17α-ol-3,11,20-trione, 4-Pregnene-17α,21-diol-3,11,20-trione 21-acetate
Empirical Formula (Hill Notation):
C
23
H
30
O
6
CAS Number:
50-04-4
Molecular Weight:
402.48
EC Number:
200-006-5
Products
Product Category
bioactive small molecules (1)
hormones (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Sterility
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C3130
Cortisone 21-acetate,
≥99%
Expand
View Pricing