Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
pyrex
®
media bottle
Products
Product Category
bottles (5)
Manufacturer
Capacity (L)
Capacity (mL)
Height (mm)
OD (mm)
Cap
Bottle material
Color
Vial Bottle Material
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS13951LHTC
Corning
®
pyrex
®
media bottle,
capacity 1 L, with PBT high temperature caps
Expand
View Pricing
CLS1395500HTC
Corning
®
pyrex
®
media bottle,
capacity 500 mL, with PBT high temperature caps
Expand
View Pricing
CLS13952LHTC
Corning
®
pyrex
®
media bottle,
capacity 2 L, with PBT high temperature caps
Expand
View Pricing
CLS13955LHTC
Corning
®
pyrex
®
media bottle,
capacity 5 L, with PBT high temperature caps
Expand
View Pricing
CLS1395250HTC
Corning
®
pyrex
®
media bottle,
capacity 250 mL, with PBT high temperature caps
Expand
View Pricing