Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Corning
®
DeckWorks
™
low binding tips
Products
Product Category
pipette tips (9)
Manufacturer
Binding Type
Volume Range (µL)
Color
Package Size
Special Grade
Natural (8)
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CLS4151
Corning
®
DeckWorks
™
low binding tips,
1 - 200μL low binding pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4154
Corning
®
DeckWorks
™
low binding tips,
1 - 200μL low binding pipet tips, graduated, reload System, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
CLS4150
Corning
®
DeckWorks
™
low binding tips,
0.1 - 10μL low binding pipet tips, extended length, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4149
Corning
®
DeckWorks
™
low binding tips,
1 - 300μL low binding pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4153
Corning
®
DeckWorks
™
low binding tips,
0.1 - 10μL low binding pipet tips, extended length, graduated, reload system, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
CLS4147
Corning
®
DeckWorks
™
low binding tips,
0.1 - 10μL low binding pipet tips, extended length, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4148
Corning
®
DeckWorks
™
low binding tips,
1 - 200μL low binding pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4152
Corning
®
DeckWorks
™
low binding tips,
in hinged racks
Expand
View Pricing
CLS4155
Corning
®
DeckWorks
™
low binding tips,
1 - 300μL low binding pipet tips, graduated, reload System, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More