![Copper(II) 2,3-naphthalocyanine Dye content 85 %](/deepweb/assets/sigmaaldrich/product/structures/539/026/0d5eaaa8-b7b9-4625-8e31-c10ead033e5a/640/0d5eaaa8-b7b9-4625-8e31-c10ead033e5a.png)
Copper(II) 2,3-naphthalocyanine
![Copper(II) 2,3-naphthalocyanine Dye content 85 %](/deepweb/assets/sigmaaldrich/product/structures/539/026/0d5eaaa8-b7b9-4625-8e31-c10ead033e5a/640/0d5eaaa8-b7b9-4625-8e31-c10ead033e5a.png)
Synonym(s):
2,3-Naphthalocyanine copper(II) salt
Empirical Formula (Hill Notation):
C48H24CuN8
CAS Number:
Molecular Weight:
776.30
Products
Brand
Formula Weight
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing