Concanavalin A from Canavalia ensiformis (Jack bean)
Synonym(s):
Con A
CAS Number:
EC Number:
Products
Brand
Biological Source
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing