Cobalt(II) sulfate heptahydrate
Synonym(s):
Cobaltous sulfate heptahydrate, Cobalt(II) sulfate heptahydrate
Linear Formula:
CoSO4 · 7H2O
CAS Number:
Molecular Weight:
281.10
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Showing 1-4 of 4