C-Methylcalix[4]resorcinarene
Empirical Formula (Hill Notation):
C32H32O8
CAS Number:
Molecular Weight:
544.59
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing