Cloxacillin Sodium
Synonym(s):
Cloxacillin sodium salt monohydrate
Empirical Formula (Hill Notation):
C19H17ClN3NaO5S · H2O
CAS Number:
Molecular Weight:
475.88
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing