
Clomipramine impurity D

Synonym(s):
3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine
Empirical Formula (Hill Notation):
C19H22Cl2N2
CAS Number:
Molecular Weight:
349.30
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing