![Citropten phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/983/765/df469a1c-fdb6-4ada-ba66-7fe4cc570df8/640/df469a1c-fdb6-4ada-ba66-7fe4cc570df8.png)
Citropten
![Citropten phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/983/765/df469a1c-fdb6-4ada-ba66-7fe4cc570df8/640/df469a1c-fdb6-4ada-ba66-7fe4cc570df8.png)
Synonym(s):
5,7-Dimethoxycoumarin, Citropten, Limettin
Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing