Skip to Content
Merck
Citraconic anhydride 98%

Citraconic anhydride

Citraconic anhydride 98%
Synonym(s):
Citraconic anhydride
Empirical Formula (Hill Notation):
C5H4O3
CAS Number:
Molecular Weight:
112.08

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing