Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cholic acid
Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholanic acid, Cholanic acid
Empirical Formula (Hill Notation):
C
24
H
40
O
5
CAS Number:
81-25-4
Molecular Weight:
408.57
EC Number:
201-337-8
Products
Product Category
detergents (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Functional Group
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (3)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C1129
Cholic acid,
from bovine and/or ovine, ≥98%
Expand
View Pricing
1133503
Cholic acid,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
C2158000
Cholic acid,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP618
Cholic acid,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing