Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Chlorotrimethylsilane
Synonym(s):
TMCS, Trimethylchlorosilane, Trimethylsilyl chloride, Chlorotrimethylsilane
Linear Formula:
(CH
3
)
3
SiCl
CAS Number:
75-77-4
Molecular Weight:
108.64
EC Number:
200-900-5
Products
Brand
Sigma-Aldrich (5)
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Reaction Type
Reagent Type
Special Grade
Puriss (1)
Purum (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
92361
Chlorotrimethylsilane,
≥98.0% (GC)
Expand
View Pricing
386529
Chlorotrimethylsilane,
purified by redistillation, ≥99%
Expand
View Pricing
8.18737
Chlorotrimethylsilane,
for synthesis
Expand
View Pricing
95541
Chlorotrimethylsilane,
Wacker Chemie AG, ≥99.0% (GC)
Expand
View Pricing
92360
Chlorotrimethylsilane,
puriss., ≥99.0% (GC)
Expand
View Pricing
89595
Chlorotrimethylsilane,
derivatization grade (GC derivatization), LiChropur
™
, ≥99.0% (GC)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More