Skip to Content
Merck
Chloro(dodecyl)dimethylsilane ≥95.0% (GC)

Chloro(dodecyl)dimethylsilane

Chloro(dodecyl)dimethylsilane ≥95.0% (GC)
Synonym(s):
Chloro-dimethyl-dodecylsilane, Dimethyldodecylchlorosilane, Dodecyldimethylchlorosilane
Linear Formula:
H3C(CH2)11Si(CH3)2Cl
CAS Number:
Molecular Weight:
262.93
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing