Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorocholine chloride
Synonym(s):
(2-Chloroethyl)trimethylammonium chloride, Chlormequat chloride, Choline dichloride
Linear Formula:
(CH
3
)
3
N(Cl)CH
2
CH
2
Cl
CAS Number:
999-81-5
Molecular Weight:
158.07
EC Number:
213-666-4
Products
Formula Weight
Physical Form
Product Line
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C4049
Chlorocholine chloride,
suitable for plant cell culture, BioReagent
Expand
View Pricing