Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorobenzene
Synonym(s):
Chlorobenzene
Empirical Formula (Hill Notation):
C
6
H
5
Cl
CAS Number:
108-90-7
Molecular Weight:
112.56
EC Number:
203-628-5
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (7)
Supelco (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (2)
ACS reagent (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
101389
Chlorobenzene,
ReagentPlus
®
, 99%
Expand
View Pricing
319996
Chlorobenzene,
ACS reagent, ≥99.5%
Expand
View Pricing
284513
Chlorobenzene,
anhydrous, 99.8%
Expand
View Pricing
270644
Chlorobenzene,
suitable for HPLC, 99.9%
Expand
View Pricing
8.01791
Chlorobenzene,
for synthesis
Expand
View Pricing
08650
Chlorobenzene,
analytical standard
Expand
View Pricing
8.14506
Chlorobenzene,
special quality for synthesis
Expand
View Pricing
PHR1665
Chlorobenzene,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
CX0855
Chlorobenzene,
Meets ACS Specifications GR ACS
Expand
View Pricing
900521
Chlorobenzene,
anhydrous, ZerO
2
®
, 99.8%
Expand
View Pricing