![Cephalexin Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/341/314/1f186ca8-a58c-44a5-bab3-f50df4d25e26/640/1f186ca8-a58c-44a5-bab3-f50df4d25e26.jpg)
Cephalexin
![Cephalexin Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/341/314/1f186ca8-a58c-44a5-bab3-f50df4d25e26/640/1f186ca8-a58c-44a5-bab3-f50df4d25e26.jpg)
Synonym(s):
Cephalexin monohydrate
Empirical Formula (Hill Notation):
C16H17N3O4S · H2O
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing