Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cefixime trihydrate
Empirical Formula (Hill Notation):
C
16
H
15
N
5
O
7
S
2
· 3H
2
O
CAS Number:
125110-14-7
Molecular Weight:
507.50
Products
Brand
Supelco (2)
Formula Weight
Purity
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
18588
Cefixime trihydrate,
suitable for microbiology, ≥98.0% (HPLC)
Expand
View Pricing
PHR3833
Cefixime trihydrate,
certified reference material, pharmaceutical secondary standard
Expand
View Pricing