Skip to Content
Merck
Carvedilol British Pharmacopoeia (BP) Reference Standard

Carvedilol

Carvedilol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190, Carvedilol
Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique