Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon - vitreous - 1000c
Products
Manufacturer
Physical Form
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GF81202755
Carbon - vitreous - 1000c,
foil, thickness 0.10 mm, size 50 × 50 mm, tolerance +0/-0.01mm
Expand
View Pricing
GF71432917
Carbon - vitreous - 1000c,
foil, thickness 0.06 mm, size 50 × 50 mm, tolerance +0/-0.01mm
Expand
View Pricing
GF98645338
Carbon - vitreous - 1000c,
foil, thickness 0.10 mm, size 100 × 100 mm, tolerance +0/-0.01mm
Expand
View Pricing