Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon nanotube forestvertically aligned on Si substrate
Synonym(s):
CNT array, MWCNT, MWCNT array
CAS Number:
308068-56-6
Products
Brand
Sigma-Aldrich (2)
Purity
Height (mm)
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
901046
Carbon nanotube forestvertically aligned on Si substrate,
Drawable, DxL 10-40 nm× 1.0 mm ± 0.5 mm
Expand
View Pricing
901051
Carbon nanotube forestvertically aligned on Si substrate,
DxL10-40 nm × 0.5 ± 0.1 mm
Expand
View Pricing