![Candesartan cilexetil European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/764/072/65be62f7-edd9-4bb9-bc9c-4e8832cab1dc/640/65be62f7-edd9-4bb9-bc9c-4e8832cab1dc.png)
Candesartan cilexetil
![Candesartan cilexetil European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/764/072/65be62f7-edd9-4bb9-bc9c-4e8832cab1dc/640/65be62f7-edd9-4bb9-bc9c-4e8832cab1dc.png)
Synonym(s):
2-ethoxy-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-Benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, TCV 116, TCY 116, Candesartan cilexetil
Empirical Formula (Hill Notation):
C33H34N6O6
CAS Number:
Molecular Weight:
610.66
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing